N,O-Di-boc-hydroxylamine ≥98.0% (by GC)

Supplier: TCI America
Warning

B4780-1G B4780-5G
TCB4780-1GEA 44.24 CAD
TCB4780-1G TCB4780-5G
N,O-Di-boc-hydroxylamine ≥98.0% (by GC)
N,O-Di-boc-hydroxylamine
Formula: C₁₀H₁₉NO₅
Storage Temperature: Freezer
MDL Number: MFCD00034797
CAS Number: 85006-25-3
EINECS: 285-055-0

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 67.0 to 71.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR