Methyl (R)-(+)-3-boc-2,2-dimethyl-4-oxazolidinecarboxylate ≥96.0% (by GC)
Supplier: TCI America
|
Warning
|
B5352-1G
B5352-5G
76219-586EA
186.44
CAD
76219-586
76219-588
Methyl (R)-(+)-3-boc-2,2-dimethyl-4-oxazolidinecarboxylate ≥96.0% (by GC)
Methyl (R)-(+)-3-boc-2,2-dimethyl-4-oxazolidinecarboxylate
Formula:
C₁₂H₂₁NO₅ Boiling Pt: 102°C Flash Pt: 93°C |
MDL Number:
MFCD00674041 CAS Number: 95715-86-9 |
Specification Test Results
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 96.0 % |
Optical purity (GC) | min. 96.0 ee% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...