Isopropyl methanesulfonate ≥98.0% (by GC)

Supplier: TCI America
Danger

I0914-1G I0914-5G
TCI0914-1GEA 66.04 CAD
TCI0914-1G TCI0914-5G
Isopropyl methanesulfonate ≥98.0% (by GC)
Isopropyl methanesulfonate
Formula: C₄H₁₀O₃S
Boiling Pt: 67°C
Melting Pt: 7°C
Flash Pt: 104°C
MDL Number: MFCD00047802
CAS Number: 926-06-7
EINECS: 213-132-0
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR