(R)-N-Fmoc-(2-furyl)alanine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

F0934-1G F0934-200MG
TCF0934-1GEA 331.8 CAD
TCF0934-1G TCF0934-200MG
(R)-N-Fmoc-(2-furyl)alanine ≥98.0% (by HPLC, titration analysis)
(R)-N-Fmoc-(2-furyl)alanine
Formula: C₂₂H₁₉NO₅
MDL Number: MFCD01311774
CAS Number: 220497-85-8

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 146.0 to 150.0 deg-C
Specific rotation [a]20/D +33.0 to +37.0 deg (C=1,DMF)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR