Methyl-2-(3-fluorophenyl)acetate ≥98.0% (by GC)

Supplier: TCI America

M2536-25G M2536-5G
TCM2536-25GEA 343.81 CAD
TCM2536-25G TCM2536-5G
Methyl-2-(3-fluorophenyl)acetate ≥98.0% (by GC)
Methyl-2-(3-fluorophenyl)acetate
Formula: C₉H₉FO₂
MDL Number: MFCD03093288
CAS Number: 64123-77-9

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR