1-Hydroxypyrene 98,0 GC_ASSAY_METHOD

Supplier: TCI America
Warning

H1435-1G H1435-5G
TCH1435-1GEA 308.1 CAD
TCH1435-1G TCH1435-5G
1-Hydroxypyrene 98,0 GC_ASSAY_METHOD
1-Hydroxypyrene
Formula: C₁₆H₁₀O
MW: 218.255 g/mol
MDL Number: MFCD00044543
CAS Number: 5315-79-7

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR