Ethyl-1-bromocyclobutanecarboxylate ≥96.0% (by GC)
Supplier: TCI America
|
Danger
|
E1245-1G
E1245-5G
TCE1245-1GEA
67.94
CAD
TCE1245-1G
TCE1245-5G
Ethyl-1-bromocyclobutanecarboxylate ≥96.0% (by GC)
Ethyl-1-bromocyclobutanecarboxylate
Formula:
C₇H₁₁BrO₂ Boiling Pt: 88°C Flash Pt: 79°C Storage Temperature: Refrigerator |
MDL Number:
MFCD00001322 CAS Number: 35120-18-4 EINECS: 252-384-6 UN: 3265 ADR: 8,II |
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...