3-Phenoxypropan-1-ol ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
3-Phenoxy-1-propanol
P2254-25G
P2254-5G
TCP2254-25GEA
277.76
CAD
TCP2254-25G
TCP2254-5G
3-Phenoxypropan-1-ol ≥98.0% (by GC)
3-Phenoxypropan-1-ol
Formula:
C₉H₁₂O₂ Boiling Pt: 153°C |
MDL Number:
MFCD00039549 CAS Number: 6180-61-6 EINECS: 228-228-8 |
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...