Diethyl-trans-1,2-cyclopropanedicarboxylate ≥96.0% (by GC)

Supplier: TCI America

D4743-1G D4743-5G
TCD4743-1GEA 48.98 CAD
TCD4743-1G TCD4743-5G
Diethyl-trans-1,2-cyclopropanedicarboxylate ≥96.0% (by GC)
Diethyl-trans-1,2-cyclopropanedicarboxylate
Formula: C₉H₁₄O₄
Boiling Pt: 71°C
Flash Pt: 98°C
MDL Number: MFCD00198002
CAS Number: 3999-55-1

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR