1-Phenylcyclobutanecarbonitrile ≥98.0% (by GC)

Supplier: TCI America
Warning

P2252-25G P2252-5G
TCP2252-25GEA 358.66 CAD
TCP2252-25G TCP2252-5G
1-Phenylcyclobutanecarbonitrile ≥98.0% (by GC)
1-Phenylcyclobutanecarbonitrile
Formula: C₁₁H₁₁N
MW: 157.216 g/mol
Boiling Pt: 85°C
MDL Number: MFCD00019258
CAS Number: 14377-68-5
EINECS: 238-351-9

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR