2-Phenylacetohydrazide ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

P2203-25G P2203-5G
TCP2203-25GEA 442.4 CAD
TCP2203-25G TCP2203-5G
2-Phenylacetohydrazide ≥98.0% (by GC, titration analysis)
2-Phenylacetohydrazide
Formula: C₈H₁₀N₂O
MDL Number: MFCD00007612
CAS Number: 937-39-3

Order Now

Specification Test Results

Appearance White to Gray to Red powder to crystal
Purity (GC) min. 98.0 %
Purity (Bromination Method) min. 98.0 %
Melting point 115.0 to 119.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR