1-(cyclopropylcarbonyl)piperazine hydrochloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

Synonyms: Cyclopropyl(piperazin-1-yl)methanone hydrochloride, 1-(Cyclopropylcarbonyl)piperazine hydrochloride

C3112-1G C3112-5G
TCC3112-1GEA 93.22 CAD
TCC3112-1G TCC3112-5G
1-(cyclopropylcarbonyl)piperazine hydrochloride ≥98.0% (by titrimetric analysis)
1-(cyclopropylcarbonyl)piperazine hydrochloride
Formula: C₈H₁₅ClN₂O
MW: 190.671 g/mol
MDL Number: MFCD03428576
CAS Number: 1021298-67-8

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (Argentmetric Titration) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 175.0 to 179.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR