1,3-Dibromo-4-nitrobenzene ≥98.0% (by GC)

Supplier: TCI America
Warning

D4790-1G D4790-5G
TCD4790-1GEA 37.92 CAD
TCD4790-1G TCD4790-5G
1,3-Dibromo-4-nitrobenzene ≥98.0% (by GC)
1,3-Dibromo-4-nitrobenzene
Formula: C₆H₃Br₂NO₂
MDL Number: MFCD02660416
CAS Number: 51686-78-3

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 59.0 to 63.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR