n-Dodecyl acrylate ≥98.0% (by GC) stabilised

Supplier: TCI America
Warning

Synonyms: Acrylic acid Lauryl ester, Acrylic acid Dodecyl ester, Dodecyl Acrylate, Lauryl Acrylate

Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

D4129-100ML D4129-25ML D4129-500ML
TCD4129-100MLEA 100.71 CAD
TCD4129-100ML TCD4129-25ML TCD4129-500ML
n-Dodecyl acrylate ≥98.0% (by GC) stabilised
n-Dodecyl acrylate
Formula: C₁₅H₂₈O₂
Boiling Pt: 120°C
Melting Pt: 4°C
Flash Pt: 145°C
MDL Number: MFCD00042871
CAS Number: 2156-97-0
EINECS: 218-463-4

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR