3,5-Bis(methoxycarbonyl)benzeneboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1,3-benzenedicarboxylic acid dimethyl ester, Dimethyl 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)isophthalate, 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)isophthalic acid dimethyl ester, 3,5-Bis(methoxycarbonyl)benzeneboronic acid pinacol ester, Dimethyl 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1,3-benzenedicarboxylate, 2-[3,5-Bis(methoxycarbonyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
D5239-1G
D5239-5G
TCD5239-1GEA
210.14
CAD
TCD5239-1G
TCD5239-5G
3,5-Bis(methoxycarbonyl)benzeneboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
3,5-Bis(methoxycarbonyl)benzeneboronic acid pinacol ester
Formula:
C₁₆H₂₁BO₆ MW: 320.148 g/mol |
MDL Number:
MFCD11858596 CAS Number: 944392-68-1 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 135.0 to 139.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...