Ethyl potassium oxalate ≥95.0% (by titrimetric analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
Potassium 2-ethoxy-2-oxoacetate
E1184-25G
E1184-5G
TCE1184-25GEA
706.26
CAD
TCE1184-25G
TCE1184-5G
Ethyl potassium oxalate ≥95.0% (by titrimetric analysis)
Ethyl potassium oxalate
Formula:
C₄H₅KO₄ MW: 156.178 g/mol |
MDL Number:
MFCD00045861 CAS Number: 1906-57-6 EINECS: 217-606-8 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (Nonaqueous Titration) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...