6-Hydroxy-1,2,3,4-tetrahydroisoquinoline ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
H1572-1G
H1572-5G
TCH1572-1GEA
246.48
CAD
TCH1572-1G
TCH1572-5G
6-Hydroxy-1,2,3,4-tetrahydroisoquinoline ≥98.0% (by GC)
6-Hydroxy-1,2,3,4-tetrahydroisoquinoline
Formula:
C₉H₁₁NO |
MDL Number:
MFCD01717047 CAS Number: 14446-24-3 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 96.0 % |
Melting point | 190.0 to 194.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...