N-Boc-2-bromobenzylamine ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: N-Boc-2-bromobenzylamine, 2-Bromo-N-(tert-butoxycarbonyl)benzylamine, tert-Butyl (2-Bromobenzyl)carbamate, (2-Bromobenzyl)carbamic acid tert-Butyl ester

B4920-1G B4920-5G
TCB4920-1GEA 94.8 CAD
TCB4920-1G TCB4920-5G
N-Boc-2-bromobenzylamine ≥98.0% (by GC)
N-Boc-2-bromobenzylamine
Formula: C₁₂H₁₆BrNO₂
MW: 286.169 g/mol
MDL Number: MFCD05664385
CAS Number: 162356-90-3

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 51.0 to 55.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR