5-Bromo-4-chloro-3-indolyl-β-D-glucoside ≥98.0% (by HPLC)
Supplier: TCI America
B5393-1G
B5393-200MG
TCB5393-1GEA
156.42
CAD
TCB5393-1G
TCB5393-200MG
5-Bromo-4-chloro-3-indolyl-β-D-glucoside ≥98.0% (by HPLC)
5-Bromo-4-chloro-3-indolyl-beta-D-glucoside
Formula:
C₁₄H₁₅BrClNO₆ Melting Pt: 243°C Storage Temperature: Freezer |
MDL Number:
MFCD00063690 CAS Number: 15548-60-4 EINECS: 239-603-0 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 97.0 % |
Specific rotation [a]20/D | -78.0 to -83.0 deg (C=0.5,DMF) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...