(9-Ethyl-9H-carbazol-3-yl)boronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
|
Warning
|
E1153-1G
E1153-5G
TCE1153-1GEA
88.48
CAD
TCE1153-1G
TCE1153-5G
(9-Ethyl-9H-carbazol-3-yl)boronic acid (contains varying amounts of Anhydride)
(9-Ethyl-9H-carbazol-3-yl)boronic acid
Formula:
C₁₄H₁₄BNO₂ |
MDL Number:
MFCD09743121 CAS Number: 669072-93-9 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | 97.0 to 109.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...