2-(4-Bromophenyl)propan-2-ol ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 2-(4-Bromophenyl)-2-propanol

B5274-1G B5274-5G
76219-552EA 191.18 CAD
76219-552 76219-554
2-(4-Bromophenyl)propan-2-ol ≥98.0% (by GC)
2-(4-Bromophenyl)propan-2-ol
Formula: C₉H₁₁BrO
Boiling Pt: 85°C
Storage Temperature: Refrigerator
MDL Number: MFCD11870096
CAS Number: 2077-19-2

Order Now

Specification Test Results

Appearance White to Orange to Green powder to lump
Purity (GC) min. 98.0 %
Melting point 42.0 to 46.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR