2-Bromo-4'-methylpropiophenone ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
B4781-25G
B4781-5G
TCB4781-25GEA
259.44
CAD
TCB4781-25G
TCB4781-5G
2-Bromo-4'-methylpropiophenone ≥98.0% (by GC)
2-Bromo-4'-methylpropiophenone
Formula:
C₁₀H₁₁BrO MW: 227.101 g/mol Boiling Pt: 162°C |
MDL Number:
MFCD11131402 CAS Number: 1451-82-7 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 78.0 to 82.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...