1-bromo-4-iodonaphthalene ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
B4864-1G
B4864-5G
76219-436EA
116.92
CAD
76219-436
76219-438
1-bromo-4-iodonaphthalene ≥98.0% (by GC)
1-bromo-4-iodonaphthalene
Formula:
C₁₀H₆BrI |
MDL Number:
MFCD01166242 CAS Number: 63279-58-3 |
Specification Test Results
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 88.0 to 92.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...