3-Bromo-2-fluorophenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

B5137-1G B5137-5G
TCB5137-1GEA 48.98 CAD
TCB5137-1G TCB5137-5G
3-Bromo-2-fluorophenylboronic acid (contains varying amounts of Anhydride)
3-Bromo-2-fluorophenylboronic acid
Formula: C₆H₅BBrFO₂
MDL Number: MFCD05664228
CAS Number: 352535-97-8

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) 97.0 to 109.0 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR