3-Hydroxymethyl-4-methoxybenzeneboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

H1563-1G H1563-5G
TCH1563-1GEA 83.74 CAD
TCH1563-1G TCH1563-5G
3-Hydroxymethyl-4-methoxybenzeneboronic acid (contains varying amounts of Anhydride)
3-Hydroxymethyl-4-methoxybenzeneboronic acid
Formula: C₈H₁₁BO₄
MDL Number: MFCD14155851
CAS Number: 908142-03-0

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (Neutralization titration) 97.0 to 111.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR