tert-Butyl hydrogen succinate ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
M3033-1G
M3033-5G
TCM3033-1GEA
37.92
CAD
TCM3033-1G
TCM3033-5G
tert-Butyl hydrogen succinate ≥98.0% (by GC, titration analysis)
tert-Butyl hydrogen succinate
Formula:
C₈H₁₄O₄ Boiling Pt: 92°C |
MDL Number:
MFCD00273441 CAS Number: 15026-17-2 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 51.0 to 55.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...