4-(9-Carbazolyl)benzeneboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

C2926-1G C2926-5G
TCC2926-1GEA 118.97 CAD
TCC2926-1G TCC2926-5G
4-(9-Carbazolyl)benzeneboronic acid (contains varying amounts of Anhydride)
4-(9-Carbazolyl)benzeneboronic acid
Formula: C₁₈H₁₄BNO₂
Melting Pt: 264°C
MDL Number: MFCD13176534
CAS Number: 419536-33-7

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 107.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR