4-methoxy-1-naphthoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 4-Methoxynaphthalene-1-carboxylic acid, 4-Methoxy-1-naphthoic acid

M2763-1G M2763-5G
TCM2763-1GEA 153.26 CAD
TCM2763-1G TCM2763-5G
4-methoxy-1-naphthoic acid ≥98.0% (by GC, titration analysis)
4-methoxy-1-naphthoic acid
Formula: C₁₂H₁₀O₃
MW: 202.209 g/mol
MDL Number: MFCD02575372
CAS Number: 13041-62-8

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 243.0 to 247.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR