2-(3-Bromophenyl)naphthalene ≥95.0% (by GC)

Supplier: TCI America

B4568-1G B4568-5G
TCB4568-1GEA 53.72 CAD
TCB4568-1G TCB4568-5G
2-(3-Bromophenyl)naphthalene ≥95.0% (by GC)
2-(3-Bromophenyl)naphthalene
Formula: C₁₆H₁₁Br
MDL Number: MFCD16658911
CAS Number: 667940-23-0

Order Now

Specification Test Results

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 95.0 %
Melting point 97.0 to 101.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR