2-Chloropyrimidine-5-boronic acid pinacol ester ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 2-chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidine, 2-chloro-5-(tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidine

C3436-1G C3436-5G
TCC3436-1GEA 120.08 CAD
TCC3436-1G TCC3436-5G
2-Chloropyrimidine-5-boronic acid pinacol ester ≥98.0% (by GC, titration analysis)
2-Chloropyrimidine-5-boronic acid pinacol ester
Formula: C₁₀H₁₄BClN₂O₂
MW: 240.494 g/mol
MDL Number: MFCD11856048
CAS Number: 1003845-08-6

Order Now

Specification Test Results

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 65.0 to 69.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR