5-Bromo-2,4-dimethoxybenzoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

B5192-1G B5192-5G
TCB5192-1GEA 82.16 CAD
TCB5192-1G TCB5192-5G
5-Bromo-2,4-dimethoxybenzoic acid ≥98.0% (by GC, titration analysis)
5-Bromo-2,4-dimethoxybenzoic acid
Formula: C₉H₉BrO₄
MW: 261.071 g/mol
MDL Number: MFCD05865172
CAS Number: 32246-20-1

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR