1H-1,2,4-Triazole-1-carboxamidine hydrochloride ≥98.0% (by titrimetric analysis)
Supplier: TCI America
|
Danger
|
T3124-1G
T3124-5G
TCT3124-1GEA
26.86
CAD
TCT3124-1G
TCT3124-5G
1H-1,2,4-Triazole-1-carboxamidine hydrochloride ≥98.0% (by titrimetric analysis)
1H-1,2,4-Triazole-1-carboxamidine hydrochloride
Formula:
C₃H₆ClN₅ Melting Pt: 199°C |
MDL Number:
MFCD03095468 CAS Number: 19503-26-5 UN: 2811 ADR: 6.1,III |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...