Violanthrone-79 ≥95.0% (by HPLC)

Supplier: TCI America

V0131-1G V0131-5G
TCV0131-1GEA 271.76 CAD
TCV0131-1G TCV0131-5G
Violanthrone-79 ≥95.0% (by HPLC)
Violanthrone-79
Formula: C₅₀H₄₈O₄
MDL Number: MFCD00491570
CAS Number: 85652-50-2

Order Now

Specification Test Results

Appearance Orange to Brown to Dark purple powder to crystal
Purity (HPLC) min. 95.0 area%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR