4-[Difluoro(3,4,5-trifluorophenoxy)methyl]-3,5-difluoro-4'-n-propylbiphenyl ≥98.0% (by GC)

Supplier: TCI America

D5130-1G D5130-5G
TCD5130-1GEA 129.56 CAD
TCD5130-1G TCD5130-5G
4-[Difluoro(3,4,5-trifluorophenoxy)methyl]-3,5-difluoro-4'-n-propylbiphenyl ≥98.0% (by GC)
4-[Difluoro(3,4,5-trifluorophenoxy)methyl]-3,5-difluoro-4'-n-propylbiphenyl
Formula: C₂₂H₁₅F₇O
Melting Pt: 49°C
MDL Number: MFCD18451990
CAS Number: 303186-20-1

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR