(1R,2R,3S,5R)-(-)-2,3-Pinanediol ≥98.0% (by GC)
Supplier: TCI America
P2245-1G
P2245-5G
TCP2245-1GEA
60.04
CAD
TCP2245-1G
TCP2245-5G
(1R,2R,3S,5R)-(-)-2,3-Pinanediol ≥98.0% (by GC)
(1R,2R,3S,5R)-(-)-2,3-Pinanediol
Formula:
C₁₀H₁₈O₂ Boiling Pt: 90°C |
MDL Number:
MFCD09955216 CAS Number: 22422-34-0 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 54.0 to 58.0 deg-C |
Specific rotation [a]20/D | -8.0 to -11.0 deg (C=6.5, Toluene) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...