1,1-Bis(hydroxymethyl)cyclopropane ≥96.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 1,1-Bis(Hydroxymethyl)cyclopropane

B5365-25G B5365-5G
76219-598EA 90.06 CAD
76219-598 76219-600
1,1-Bis(hydroxymethyl)cyclopropane ≥96.0% (by GC)
1,1-Bis(hydroxymethyl)cyclopropane
Formula: C₅H₁₀O₂
Boiling Pt: 98°C
MDL Number: MFCD00464207
CAS Number: 39590-81-3

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR