5-Chloro-2-phenylimidazole-4-carboxaldehyde ≥97.0% (by HPLC, total nitrogen)

Supplier: TCI America
Warning

C3018-1G C3018-5G
TCC3018-1GEA 76 CAD
TCC3018-1G TCC3018-5G
5-Chloro-2-phenylimidazole-4-carboxaldehyde ≥97.0% (by HPLC, total nitrogen)
5-Chloro-2-phenylimidazole-4-carboxaldehyde
Formula: C₁₀H₇ClN₂O
MDL Number: MFCD01935953
CAS Number: 60367-52-4

Order Now

Specification Test Results

Appearance Orange to Amber to Dark red powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (with Total Nitrogen) min. 97.0 %
Melting point 169.0 to 173.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR