1-Boc-3-fluoro-4-piperidone ≥97.0% (by GC)

Supplier: TCI America
Warning

B4858-1G B4858-200MG
76219-432EA 496.12 CAD
76219-432 76219-434
1-Boc-3-fluoro-4-piperidone ≥97.0% (by GC)
1-Boc-3-fluoro-4-piperidone
Formula: C₁₀H₁₆FNO₃
MDL Number: MFCD10566554
CAS Number: 211108-50-8

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 97.0 %
Melting point 74.0 to 78.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR