Phenyl acrylate ≥98.0% (by GC) stabilized

Supplier: TCI America
Warning


Stabilization: Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)

P2268-25G P2268-5G
TCP2268-25GEA 411.45 CAD
TCP2268-25G TCP2268-5G
Phenyl acrylate ≥98.0% (by GC) stabilized
Phenyl acrylate
Formula: C₉H₈O₂
Boiling Pt: 98°C
MDL Number: MFCD00048145
CAS Number: 937-41-7
EINECS: 213-329-1

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR