1H,1H,2H,2H-Tridecafluoro-n-octyl acrylate ≥98.0% (by GC) stabilised
Supplier: TCI America
|
Warning
|
Synonyms:
3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluorooctyl acrylate, Acrylic acid 1H,1H,2H,2H-Tridecafluoro-n-octyl ester
Stabilization:
Stabilized with HQ + MEHQ
T3451-25G
T3451-5G
76220-650EA
235.42
CAD
76220-650
76220-652
1H,1H,2H,2H-Tridecafluoro-n-octyl acrylate ≥98.0% (by GC) stabilised
1H,1H,2H,2H-Tridecafluoro-n-octyl acrylate
Formula:
C₁₁H₇F₁₃O₂ Boiling Pt: 50°C Flash Pt: 102°C |
MDL Number:
MFCD00042351 CAS Number: 17527-29-6 EINECS: 241-527-8 |
Specification Test Results
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...