Pemirolast potassium ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

P2547-1G P2547-5G
TCP2547-1GEA 263.86 CAD
TCP2547-1G TCP2547-5G
Pemirolast potassium ≥98.0% (by HPLC, titration analysis)
Pemirolast potassium
Formula: C₁₀H₇KN₆O
Melting Pt: 317°C
Storage Temperature: Refrigerator
MDL Number: MFCD01690051
CAS Number: 100299-08-9
Merck Index: 14,07078

Order Now

Specification Test Results

Appearance Light yellow to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR