Sodium methacrylate ≥98.0% (by titrimetric analysis) stabilised

Supplier: TCI America
Warning


Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

S0987-100G S0987-25G
TCS0987-100GEA 195.92 CAD
TCS0987-100G TCS0987-25G
Sodium methacrylate ≥98.0% (by titrimetric analysis) stabilised
Sodium methacrylate
Formula: C₄H₅NaO₂
MW: 108.072 g/mol
MDL Number: MFCD00045886
CAS Number: 5536-61-8
EINECS: 226-896-5

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Bromination Method) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR