3,4-Difluoro-4'-(4-ethylcyclohexyl)biphenyl ≥98.0% (by GC)

Supplier: TCI America

D4859-25G D4859-5G
TCD4859-25GEA 755.4 CAD
TCD4859-25G TCD4859-5G
3,4-Difluoro-4'-(4-ethylcyclohexyl)biphenyl ≥98.0% (by GC)
3,4-Difluoro-4'-(4-ethylcyclohexyl)biphenyl
Formula: C₂₀H₂₂F₂
MDL Number: MFCD12828121
CAS Number: 134412-18-3

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 68.0 to 72.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR