trans,trans-4-(3,4-Difluorophenyl)-4'-n-propylbicyclohexyl ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
trans,trans-4-(3,4-Difluorophenyl)-4'-propylbicyclohexyl, trans,trans-4-(3,4-Difluorophenyl)-4'-propyl-1,1'-bi(cyclohexane)
D4797-1G
D4797-5G
TCD4797-1GEA
63.2
CAD
TCD4797-1G
TCD4797-5G
trans,trans-4-(3,4-Difluorophenyl)-4'-n-propylbicyclohexyl ≥98.0% (by GC)
trans,trans-4-(3,4-Difluorophenyl)-4'-n-propylbicyclohexyl
Formula:
C₂₁H₃₀F₂ |
MDL Number:
MFCD09838997 CAS Number: 82832-57-3 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 42.0 to 46.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...