trans-4-Ethyl-4'-(4-n-pentylcyclohexyl)biphenyl ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 4-Ethyl-4'-(trans-4-pentylcyclohexyl)biphenyl, 4-Ethyl-4'-(trans-4-pentylcyclohexyl)-1,1'-biphenyl

E1254-1G E1254-5G
TCE1254-1GEA 75.84 CAD
TCE1254-1G TCE1254-5G
trans-4-Ethyl-4'-(4-n-pentylcyclohexyl)biphenyl ≥98.0% (by GC)
trans-4-Ethyl-4'-(4-n-pentylcyclohexyl)biphenyl
Formula: C₂₅H₃₄
MW: 334.547 g/mol
MDL Number: MFCD01941072
CAS Number: 79709-85-6
EINECS: 279-240-5

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Mesomorphic range 144.0 to 167.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR