trans,trans-4-(3,4-Difluorophenyl)-4'-vinylbicyclohexyl ≥98.0% (by GC)
Supplier: TCI America
D4872-1G
D4872-5G
TCD4872-1GEA
52.93
CAD
TCD4872-1G
TCD4872-5G
trans,trans-4-(3,4-Difluorophenyl)-4'-vinylbicyclohexyl ≥98.0% (by GC)
trans,trans-4-(3,4-Difluorophenyl)-4'-vinylbicyclohexyl
Formula:
C₂₀H₂₆F₂ |
MDL Number:
MFCD09842838 CAS Number: 142400-92-8 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 44.0 to 48.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...