trans,trans-4-(3,4-Difluorophenyl)-4'-n-pentylbicyclohexyl ≥98.0% (by GC)
Supplier: TCI America
D4798-1G
D4798-5G
TCD4798-1GEA
51.19
CAD
TCD4798-1G
TCD4798-5G
trans,trans-4-(3,4-Difluorophenyl)-4'-n-pentylbicyclohexyl ≥98.0% (by GC)
trans,trans-4-(3,4-Difluorophenyl)-4'-n-pentylbicyclohexyl
Formula:
C₂₃H₃₄F₂ |
MDL Number:
MFCD13182310 CAS Number: 118164-51-5 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 46.0 to 50.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...