2,4,6-Triisopropylbenzenesulfonyl azide ≥98.0% (by HPLC)

Supplier: TCI America
Warning

T3434-1G T3434-5G
76220-626EA 55.3 CAD
76220-626 76220-628
2,4,6-Triisopropylbenzenesulfonyl azide ≥98.0% (by HPLC)
2,4,6-Triisopropylbenzenesulfonyl azide
Formula: C₁₅H₂₃N₃O₂S
Melting Pt: 43°C
Storage Temperature: Refrigerator
MDL Number: MFCD00859286
CAS Number: 36982-84-0

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 12.90 to 14.40 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR