2,7-Dihydroxy-9-fluorenone ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 2,7-Dihydroxy-9H-fluoren-9-one

D1685-25G D1685-5G
76218-978EA 214.88 CAD
76218-978 76218-980
2,7-Dihydroxy-9-fluorenone ≥98.0% (by HPLC, titration analysis)
2,7-Dihydroxy-9-fluorenone
Formula: C₁₃H₈O₃
MDL Number: MFCD00037141
CAS Number: 42523-29-5

Order Now

Specification Test Results

Appearance Light yellow to Brown powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR