Ethyl-3,5-dimethylisoxazole-4-carboxylate ≥98.0% (by GC)
Supplier: TCI America
E1212-1G
E1212-5G
TCE1212-1GEA
52.14
CAD
TCE1212-1G
TCE1212-5G
Ethyl-3,5-dimethylisoxazole-4-carboxylate ≥98.0% (by GC)
Ethyl-3,5-dimethylisoxazole-4-carboxylate
Formula:
C₈H₁₁NO₃ Boiling Pt: 220°C |
MDL Number:
MFCD00052557 CAS Number: 17147-42-1 |
Specification Test Results
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...